Learn More
Kingfisher Biotech Inc Bovine IL-6 Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc KP0652B100
Encompass
The Bovine IL-6 polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Bovine IL-6 ELISA. The Bovine IL-6 antibody was produced in chickens and is antigen-affinity purified. The reactivity by species is: (Bovine IL-6 - Strong) (Canine IL-6 - None) (Caprine IL-6 - Strong) (Chicken IL-6 - None) (Cynomolgus Monkey IL-6 - None) (Dolphin IL-6 - None) (Equine IL-6 - None) (Feline IL-6 - None) (Human IL-6 - None) (Mouse IL-6 - None) (Ovine IL-6 - Strong) (Rabbit IL-6 - None) (Swine IL-6 - None) (Zebrafish IL-6 - None). For research use only. Made in the USA
Catalog No. 50-233-2330