Learn More
Kingfisher Biotech Inc Bovine IL-6 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0014B025
Encompass
The Bovine IL-6 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-6 applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IL-6 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-6 Specifications: (Molecular Weight: 20.7 kDa) (Amino Acid Sequence: GPLGEDFKND TTPGRLLLTT PEKTEALIKR MVDKISAMRK EICEKNDECE SSKETLAENK LNLPKMEEKD GCFQSGFNQA ICLIRTTAGL LEYQIYLDYL QNEYEGNQEN VRDLRKNIRT LIQILKQKIA DLITTPATNT DLLEKMQSSN EWVKNAKIIL ILRNLENFLQ FSLRAIRMK) (Gene ID: 280826). For research use only. Made in the USA
Catalog No. 50-241-3209