Learn More
Kingfisher Biotech Inc Bovine IL-8 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0023B100
Encompass
The Bovine IL-8 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine IL-8 applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine IL-8 yeast-derived recombinant protein can be purchased in multiple sizes. Bovine IL-8 Specifications: (Molecular Weight: 9.1 kDa) (Amino Acid Sequence: AVLSRMSTEL RCQCIKTHST PFHPKFIKEL RVIESGPHCE NSEIIVKLTN GNEVCLNPKE KWVQKVVQVF VKRAEKQDP) (Gene ID: 280828). For research use only. Made in the USA
Catalog No. 50-241-3505