Learn More
Cusabio Technology LLC BOVINE Immunoglobulin A ELISA
Supplier:  Cusabio Technology LLC CSBE12018B
Encompass_Preferred
The Bovine Immunoglobulin A (IgA) ELISA Kit is designed to quantitatively measure IgA levels in serum plasma cell culture supernates and tissue homogenates using a competitive ELISA method This kit detects IgA within a dynamic range of 0 39 g/mL to 25 g/mL with a sensitivity of 0 39 g/mL ensuring precise measurement of low-concentration samples The assay requires 50-100 L of sample per test and delivers results in 1-5 hours with absorbance readings taken at 450 nm Ideal for research and diagnostic applications it provides a reliable tool for studying IgA expression immune responses and disease-related biomarker analysis in bovine specimens The streamlined protocol supports flexibility in experimental workflows while maintaining robust performance
Catalog No. 50-000-48586