Learn More
Biomatik Corporation Bovine Immunoglobulin A (IgA) CLIA Kit
Supplier:  Biomatik Corporation EKU08881
Encompass_Preferred
Bovine Immunoglobulin A (IgA) CLIA Kit for detecting IgA in serum, plasma, saliva, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.04ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P40293. Gene ID: 281674. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8605