Learn More
American Research Products Inc BoVine Immunoglobulin G IgG E
Supplier:  American Research Products Inc CSBE12015B
Encompass
BoVine Immunoglobulin G IgG ELISA Kit from CUSABIO Detection Range 78 ng/ml-5000ng/ml Sensitivity 78 ng/ml Sample serum plasma Method Competitive
Catalog No. 50-003-30028