Learn More
Biomatik Corporation Bovine Immunoglobulin G, Igg Elisa Kit
Supplier:  Biomatik Corporation EKC31014
Encompass_Preferred
Bovine Immunoglobulin G, Igg Elisa Kit for detecting Immunoglobulin G, Igg in serum, plasma. Species Reactivity: Bovine (Bos taurus). Detection Range: 78-5000ng/ml. Sensitivity: 19.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5449