Learn More
American Research Products Inc BoVine Immunoglobulin M IgM E
Catalog No. 5000329050
Encompass
BoVine Immunoglobulin M IgM ELISA Kit from CUSABIO Detection Range 0 39 g/mL-25 g/mL Sensitivity 0 2 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29050 Supplier American Research Products Inc Supplier No. CSBE12017B
BoVine Immunoglobulin M IgM ELISA Kit from CUSABIO Detection Range 0 39 g/mL-25 g/mL Sensitivity 0 2 g/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive