Learn More
Biomatik Corporation Bovine Immunoglobulin M, Igm Elisa Kit
Supplier:  Biomatik Corporation EKC31015
Encompass_Preferred
Bovine Immunoglobulin M, Igm Elisa Kit for detecting Immunoglobulin M in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.39-25ug/ml. Sensitivity: 0.2ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9201