Learn More
Biomatik Corporation Bovine Inhibin B (INHB) ELISA Kit
Supplier:  Biomatik Corporation EKU04927
Encompass_Preferred
Bovine Inhibin B (INHB) ELISA Kit for detecting INHB in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 31.25-2000pg/ml. Sensitivity: 11.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42917. Target Synonyms: INH-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6221