Learn More
Biomatik Corporation Bovine Inhibin Beta A (INHbA) ELISA Kit
Supplier:  Biomatik Corporation EKU12272
Encompass_Preferred
Bovine Inhibin Beta A (INHbA) ELISA Kit for detecting INHbA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P07995. Gene ID: 281867. Target Synonyms: INH-BA; INHB-A; EDF; FRP; Activin A,Activin AB Alpha Polypeptide; Activin beta-A chain; Erythroid differentiation protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7524