Learn More
Apexbio Technology LLC Bovine insulin, 25mg, commonly used as a growth factor in cell cultures
SDP
Catalog No. NC3314006
Encompass_Preferred
Bovine insulin (insulin from the bovine pancreas), with a molecular weight of about 5800 Da, is a double-chain(α,β)peptide hormone produced in theβcells of the pancreas. Insulin is a protein hormone known to lower blood sugar, is responsible for regulating the intake of glucose, amino acids, and fatty acids by cells, and is often used as a growth supplement for cultured cells.
Catalog No. NC3314006 Supplier Apexbio Technology LLC Supplier No. A598125
Bovine insulin (insulin from the bovine pancreas), with a molecular weight of about 5800 Da, is a double-chain(α,β)peptide hormone produced in theβcells of the pancreas. Insulin is a protein hormone known to lower blood sugar, is responsible for regulating the intake of glucose, amino acids, and fatty acids by cells, and is often used as a growth supplement for cultured cells.