Learn More
Biomatik Corporation Bovine Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU05001
Encompass_Preferred
Bovine Insulin Like Growth Factor Binding Protein 4 (IGFBP4) ELISA Kit for detecting IGFBP4 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 74.07-6000pg/ml. Sensitivity: 29.62pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q05716. Gene ID: 282262. Target Synonyms: BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7010