Learn More
American Research Products Inc BoVine Insulin like growth fac
Catalog No. 5000327693
Encompass
BoVine Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-27693 Supplier American Research Products Inc Supplier No. CSBEL011088BO
BoVine Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma Method Sandwich