Learn More
American Research Products Inc BoVine Interferon y IFN y ELI
Catalog No. 5000326173
Encompass
BoVine Interferon y IFN-y ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26173 Supplier American Research Products Inc Supplier No. CSBE09812B
BoVine Interferon y IFN-y ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich