Learn More
American Research Products Inc BoVine Interleukin 1 IL 1 ELI
Catalog No. 5000330033
Encompass
BoVine Interleukin 1 IL-1 ELISA Kit from CUSABIO Detection Range 250 pg/mL-4000 pg/mL Sensitivity 250 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-30033 Supplier American Research Products Inc Supplier No. CSBE12019B
BoVine Interleukin 1 IL-1 ELISA Kit from CUSABIO Detection Range 250 pg/mL-4000 pg/mL Sensitivity 250 pg/mL Sample serum plasma tissue homogenates Method Competitive