Learn More
Biomatik Corporation Bovine Interleukin 10 (IL10) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10940
Encompass_Preferred
Bovine Interleukin 10 (IL10) High-sensitive ELISA Kit for detecting IL10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43480. Gene ID: 281246. Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5378