Learn More
American Research Products Inc BoVine Interleukin 12 IL 12 E
Catalog No. 5000326755
Encompass
BoVine Interleukin 12 IL-12 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26755 Supplier American Research Products Inc Supplier No. CSBE14010B
BoVine Interleukin 12 IL-12 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich