Learn More
Biomatik Corporation Bovine Interleukin 12B (IL12B) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10944
Encompass_Preferred
Bovine Interleukin 12B (IL12B) High-sensitive ELISA Kit for detecting IL12B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46282. Gene ID: 281857. Target Synonyms: p40; IL12-B; CLMF2; NKSF2; Natural Killer Cell Stimulatory Factor 2; Cytotoxic Lymphocyte Maturation Factor 2; Interleukin 23, P40 Subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6353