Learn More
Biomatik Corporation Bovine Interleukin 16 (IL16) ELISA Kit
Supplier:  Biomatik Corporation EKU05229
Encompass_Preferred
Bovine Interleukin 16 (IL16) ELISA Kit for detecting IL16 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q0V8R5. Gene ID: 506314. Target Synonyms: LCF; PrIL-16; Lymphocyte Chemoattractant Factor; Pro-Interleukin 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6046