Learn More
American Research Products Inc BoVine Interleukin 17A IL 17A
Catalog No. 5000328485
Encompass
BoVine Interleukin 17A (IL-17A/IL-17) ELISA Kit from CUSABIO Detection Range 0 94 pg/mL-60 pg/mL Sensitivity 0 47 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28485 Supplier American Research Products Inc Supplier No. CSBE14207B
BoVine Interleukin 17A (IL-17A/IL-17) ELISA Kit from CUSABIO Detection Range 0 94 pg/mL-60 pg/mL Sensitivity 0 47 pg/mL Sample serum plasma tissue homogenates Method Sandwich