Learn More
American Research Products Inc BoVine Interleukin 18 IL18 E
Supplier:  American Research Products Inc CSBEL011608BO
Encompass
BoVine Interleukin-18 (IL18) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27860