Learn More
American Research Products Inc BoVine Interleukin 2 IL 2 ELI
Catalog No. 5000329822
Encompass
BoVine Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 0 156ng/mL-10ng/mL Sensitivity 0 078ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29822 Supplier American Research Products Inc Supplier No. CSBE09810BIS
BoVine Interleukin 2 IL-2 ELISA Kit from CUSABIO Detection Range 0 156ng/mL-10ng/mL Sensitivity 0 078ng/ml Sample serum plasma tissue homogenates Method Sandwich