Learn More
Biomatik Corporation Bovine Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit
Supplier:  Biomatik Corporation EKU05277
Encompass_Preferred
Bovine Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit for detecting IL2Ra in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12342. Gene ID: 281861. Target Synonyms: CD25; IL2R-a; IL2-RA; IDDM10; IL2R; TCGFR; p55; TAC antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6456