Learn More
Cusabio Technology LLC BOVINE Interleukin 22 ELISA KT
Catalog No. 5000051214
Encompass_Preferred
This kit detects bovine interleukin 22 (IL22) in serum plasma and tissue homogenates using a sandwich ELISA method It provides a quantitative measurement within a detection range of 78 pg/ml to 5000 pg/ml with a sensitivity threshold of 19 5 pg/ml The assay requires 50-100 l of sample per test and delivers results within 1 to 5 hours Optical density is measured at 450 nm for accurate analyte quantification Designed for reliable performance across biological matrices this kit is suitable for researchers studying immune responses and cytokine profiling in bovine models
Catalog No. 50-000-51214 Supplier Cusabio Technology LLC Supplier No. CSBEL011635BO
This kit detects bovine interleukin 22 (IL22) in serum plasma and tissue homogenates using a sandwich ELISA method It provides a quantitative measurement within a detection range of 78 pg/ml to 5000 pg/ml with a sensitivity threshold of 19 5 pg/ml The assay requires 50-100 l of sample per test and delivers results within 1 to 5 hours Optical density is measured at 450 nm for accurate analyte quantification Designed for reliable performance across biological matrices this kit is suitable for researchers studying immune responses and cytokine profiling in bovine models