Learn More
American Research Products Inc BoVine interleukin 22 IL22 E
Catalog No. 5000327583
Encompass
BoVine interleukin 22 (IL22) ELISA kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27583 Supplier American Research Products Inc Supplier No. CSBEL011635BO
BoVine interleukin 22 (IL22) ELISA kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich