Learn More
American Research Products Inc BoVine interleukin 4 IL 4 ELI
Catalog No. 5000329801
Encompass
BoVine interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29801 Supplier American Research Products Inc Supplier No. CSBE12898B
BoVine interleukin 4 IL-4 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich