Learn More
Biomatik Corporation Bovine Interleukin 4 (IL4) CLIA Kit
Supplier:  Biomatik Corporation EKU09948
Encompass_Preferred
Bovine Interleukin 4 (IL4) CLIA Kit for detecting IL4 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.05pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P30367. Gene ID: 280824. Target Synonyms: BSF1; BCGF1 ;B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7081