Learn More
Cusabio Technology LLC Bovine interleukin 4IL-4 ELISA
Catalog No. 5000048820
Encompass_Preferred
This kit detects IL4 in bovine serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 12 5 pg/mL to 800 pg/mL with a sensitivity of 3 12 pg/mL ensuring precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours providing rapid and efficient results Measurements are performed at an absorbance wavelength of 450 nm aligning with standard microplate reader configurations Designed for reliability and ease of use this kit is suitable for researchers studying immune responses or cytokine profiling in bovine models delivering consistent performance across biological matrices
Catalog No. 50-000-48820 Supplier Cusabio Technology LLC Supplier No. CSBE12898B
This kit detects IL4 in bovine serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 12 5 pg/mL to 800 pg/mL with a sensitivity of 3 12 pg/mL ensuring precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours providing rapid and efficient results Measurements are performed at an absorbance wavelength of 450 nm aligning with standard microplate reader configurations Designed for reliability and ease of use this kit is suitable for researchers studying immune responses or cytokine profiling in bovine models delivering consistent performance across biological matrices