Learn More
American Research Products Inc BoVine Interleukin 6 IL 6 ELI
Supplier:  American Research Products Inc CSBE12899B
Encompass
BoVine Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 5 pg/mL-1000 pg/mL Sensitivity 2 5 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29950