Learn More
Biomatik Corporation Bovine Interleukin 6 (IL6) CLIA Kit
Supplier:  Biomatik Corporation EKU09910
Encompass_Preferred
Bovine Interleukin 6 (IL6) CLIA Kit for detecting IL6 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.18pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P26892. Gene ID: 280826. Target Synonyms: MGI2-A; MGI2A; HGF; BSF2; HSF; IFNB2; B-Cell Stimulatory Factor-2; Hybridoma/Plasmacytoma Growth Factor; Hepatocyte Stimulating Factor; Cytotoxic T-Cell Differentiation Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4718