Learn More
Biomatik Corporation Bovine Lactoferrin (LTF) CLIA Kit
Supplier:  Biomatik Corporation EKU08914
Encompass_Preferred
Bovine Lactoferrin (LTF) CLIA Kit for detecting LTF in serum, plasma, tissue homogenates, cell lysates, saliva, milk, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.41-300ng/ml. Sensitivity: 0.14ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P24627. Gene ID: 280846. Target Synonyms: HLF2; LF; GIG12; Lactotransferrin; Growth-inhibiting protein 12; Talalactoferrin; Lfcin-H; Kaliocin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8594