Learn More
Biomatik Corporation Bovine Lactoperoxidase (LPO) CLIA Kit
Supplier:  Biomatik Corporation EKU09787
Encompass_Preferred
Bovine Lactoperoxidase (LPO) CLIA Kit for detecting LPO in tear, milk, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P80025. Gene ID: 280844. Target Synonyms: SPO; Salivary peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5008