Learn More
American Research Products Inc BoVine lactoperoXidase LPO E
Catalog No. 5000327629
Encompass
BoVine lactoperoXidase (LPO) ELISA kit from CUSABIO Detection Range 1 56 mU/mL-100 mU/mL Sensitivity 1 56 mU/mL Sample breast milk Method Competitive
Catalog No. 50-003-27629 Supplier American Research Products Inc Supplier No. CSBEL013066BO
BoVine lactoperoXidase (LPO) ELISA kit from CUSABIO Detection Range 1 56 mU/mL-100 mU/mL Sensitivity 1 56 mU/mL Sample breast milk Method Competitive