Learn More
Biomatik Corporation Bovine Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit
Supplier:  Biomatik Corporation EKU05575
Encompass_Preferred
Bovine Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit for detecting LOX1 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P79391. Gene ID: 281368. Target Synonyms: OLR1; CLEC8A; SCARE1; Oxidized low-density lipoprotein receptor 1; C-type lectin domain family 8 member A; Lectin-like oxidized LDL receptor 1; Lectin-type oxidized LDL receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8896