Learn More
American Research Products Inc BoVine lipopolysaccharide bind
Catalog No. 5000326712
Encompass
BoVine lipopolysaccharide binding protein (LBP) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26712 Supplier American Research Products Inc Supplier No. CSBE16370B
BoVine lipopolysaccharide binding protein (LBP) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich