Learn More
American Research Products Inc Bovine LPO ELISA Kit
Catalog No. 5000459003
Encompass
This Bovine Lactoperoxidase (LPO) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in milk tears and other biological fluids This product is for Research Use Only
Catalog No. 50-004-59003 Supplier American Research Products Inc Supplier No. RDRLPOB
This Bovine Lactoperoxidase (LPO) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in milk tears and other biological fluids This product is for Research Use Only