Learn More
Kingfisher Biotech Inc Bovine M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1353B025
Encompass
The Bovine M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Bovine M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Bovine M-CSF Specifications: (Molecular Weight: 25.1 kDa) (Amino Acid Sequence: EEVSENCSHM IGNGHLLFLQ QLIDSQMETS CQISFEFVDQ EQLDDPVCYL KKAFLLVQDI MEDTMRFKDN TPNAKVIVQL QELSLRLKSC FTMDYDEQDK ACVRTFHETP LQLLEKIKNV FNETKNLLKK DWNIFSKNCN NSFAKCSSHD VVTKPDCNCL YPKATPSSDL ASVSPQQSPT PSTAPMAGLT WADSEATDGS SLLPREQALR TVDPGSAKQR PPR (223)) (Gene ID: 281094). For research use only. Made in the USA
Catalog No. 50-253-3340