Learn More
Biomatik Corporation Bovine Macrophage Derived Chemokine (MDC) ELISA Kit
Supplier:  Biomatik Corporation EKU05739
Encompass_Preferred
Bovine Macrophage Derived Chemokine (MDC) ELISA Kit for detecting MDC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A5PKF1. Gene ID: 616996. Target Synonyms: CCL22; ABCD1; DC/B-CK; SCYA22; STCP1; Chemokine C-C-Motif Ligand 22; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 22; Stimulated T-Cell Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8889