Learn More
Biomatik Corporation Bovine Matrix Metalloproteinase 2 (MMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU12304
Encompass_Preferred
Bovine Matrix Metalloproteinase 2 (MMP2) ELISA Kit for detecting MMP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9GLE5. Gene ID: 282872. Target Synonyms: CLG4; CLG4A; MMP-II; MONA; TBE-1; PEX; Gelatinase A; 72kDa Gelatinase; 72kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6354