Learn More
Biomatik Corporation Bovine Matrix Metalloproteinase 3 (MMP3) CLIA Kit
Supplier:  Biomatik Corporation EKU09586
Encompass_Preferred
Bovine Matrix Metalloproteinase 3 (MMP3) CLIA Kit for detecting MMP3 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9XSF7. Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7440