Learn More
Biomatik Corporation Bovine Matrix Metalloproteinase 13 (MMP13) CLIA Kit
Supplier:  Biomatik Corporation EKU09570
Encompass_Preferred
Bovine Matrix Metalloproteinase 13 (MMP13) CLIA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O77656. Gene ID: 281914. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8495