Learn More
Raybiotech Inc Bovine MCP-1 (CCL2) ELISA Kit
Supplier:  Raybiotech Inc ELBMCP11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.31 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Bovine MCP-1 (CCL2)
Catalog No. 50-194-8872