Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein Hydrolysate Enzymatic, Powder

Catalog No. mp210129001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210129001 1 lb.
MP210129005 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210129001 Supplier MP Biomedicals™ Supplier No. 0210129001
Only null left
Add to Cart
Add to Cart

Casein hydrolysate enzymatic is recommended for use as Animal diet

Applications:
Casein hydrolysate enzymatic is recommended for use as Animal diet.

Product Description:
Casein hydrolysate enzymatic is a pancreatic hydrolysate of casein. This hydrolysate contains, in the form of mixed amino acids and peptides, all amino acids originally present in casein. It is a refined product providing high quality soluble source of amino acids, polypeptides and peptides.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Powder
For Use With (Application) Animal diet
Quantity 1 lb.
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.