Learn More
ERIVAN BIO LLC Bovine Milk Exosomes
Catalog No. NC2613338
Encompass_Preferred
This product consists of isolated and purified exosomes derived from non-pasteurized, non-homogenized, filter-sterilized bovine milk. Milk exosomes have many benefits as bioactive ingredients due to their ability to survive gastric digestion and be taken up by intestinal cells.
Catalog No. NC2613338 Supplier ERIVAN BIO LLC Supplier No. BME 01200101
This product consists of isolated and purified exosomes derived from non-pasteurized, non-homogenized, filter-sterilized bovine milk. Milk exosomes have many benefits as bioactive ingredients due to their ability to survive gastric digestion and be taken up by intestinal cells.