Learn More
Bioss Bovine Papillomavirus
SDP
Catalog No. 5000066438
Encompass_Preferred
Bovine Papillomavirus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66438 Supplier Bioss Supplier No. BSM72021M
Bovine Papillomavirus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide