Learn More
American Research Products Inc BoVine Parathyroid hormone PT
Catalog No. 5000328209
Encompass
BoVine Parathyroid hormone (PTH) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28209 Supplier American Research Products Inc Supplier No. CSBEL018987BO
BoVine Parathyroid hormone (PTH) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich