Learn More
Biomatik Corporation Bovine Parathyroid Hormone (PTH) ELISA Kit
Supplier:  Biomatik Corporation EKU11326
Encompass_Preferred
Bovine Parathyroid Hormone (PTH) ELISA Kit for detecting PTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 9.88-800pg/ml. Sensitivity: 3.37pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01268. Gene ID: 280903. Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8167