Learn More
Kingfisher Biotech Inc Bovine PD-L1 ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY1919B004
Encompass_Preferred
The Bovine PD-L1 ELISA contains capture antibody standard and detection antibody for development of a Bovine PD-L1 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine PD-L1 ELISA can also be used to measure Bison Zebu and Wild Yak PD-L1. Alternate names include CD274 B7-H B7H1 PD-L1 PDCD1L1 PDCD1LG1 PDL1 CD274 molecule Programmed cell death 1 ligand 1. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5596