Learn More
American Research Products Inc BoVine PentraXin related prote
Catalog No. 5000328219
Encompass
BoVine PentraXin-related protein PTX3 (PTX3) ELISA kit from CUSABIO Detection Range 2 344 ng/mL-150 ng/mL Sensitivity 0 586 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28219 Supplier American Research Products Inc Supplier No. CSBEL019079BO
BoVine PentraXin-related protein PTX3 (PTX3) ELISA kit from CUSABIO Detection Range 2 344 ng/mL-150 ng/mL Sensitivity 0 586 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich